Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336920.1 | NP_001318403.1 | 3 - 4 | 1150 | 131 | |
NM_001336921.1 | NP_001324623.1 | 3 - 4 | 1150 | 131 | ||
NM_129714.4 | NP_181682.2 | 3 - 4 | 1270 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |