Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OEP16-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036450.2 | NP_001031527.1 | 1 - 2 | 719 | 66 | |
NM_001036451.2 | NP_001031528.1 | 2 - 3 | 412 | 66 | ||
NM_001036452.1 | NP_001031529.1 | 2 - 3 | 398 | 66 | ||
NM_001336959.1 | NP_001324189.1 | 2 - 3 | 612 | 66 | ||
NM_129783.4 | NP_565968.1 | 2 - 3 | 551 | 66 | ||
GenBank mRNA | AY086879.1 | - | 2 - 3 | 413 | 66 | |
BT025617.1 | - | 1 - 2 | 328 | 66 | ||
DQ386643.1 | - | 1 - 2 | 328 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |