Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129823.3 | NP_565979.1 | 1 | 2167 | 104 | |
GenBank mRNA | AF305597.1 | - | 1 | 2096 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |