Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129840.5 | NP_181807.1 | 12 - 13 | 1476 | 87 | |
GenBank mRNA | AK227008.1 | - | 12 - 13 | 1453 | 87 | |
AY048214.1 | - | 12 - 13 | 1452 | 87 | ||
AY124841.1 | - | 12 - 13 | 1394 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |