Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPK20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336999.1 | NP_001324574.1 | 5 - 6 | 1200 | 74 | |
NM_129849.5 | NP_565989.1 | 5 - 6 | 1200 | 74 | ||
GenBank mRNA | AF412082.1 | - | 5 - 6 | 1063 | 74 | |
BT001021.1 | - | 5 - 6 | 767 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |