Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084574.1 | NP_001078043.1 | 3 - 4 | 322 | 72 | |
NM_129850.4 | NP_565990.1 | 3 - 4 | 453 | 72 | ||
NM_201945.3 | NP_973674.1 | 2 - 3 | 496 | 72 | ||
GenBank mRNA | AF367316.1 | - | 3 - 4 | 279 | 72 | |
AY143940.1 | - | 3 - 4 | 195 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |