Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129863.3 | NP_181830.1 | 1 - 2 | 484 | 76 | |
GenBank mRNA | AF364952.1 | - | 1 - 2 | 402 | 76 | |
BT029025.1 | - | 1 - 2 | 42 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |