Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202813.1 | NP_001189742.1 | 3 - 4 | 374 | 108 | |
NM_129896.4 | NP_181863.1 | 3 - 4 | 382 | 108 | ||
GenBank mRNA | AY065372.1 | - | 3 - 4 | 369 | 108 | |
AY087030.1 | - | 3 - 4 | 367 | 108 | ||
AY096479.1 | - | 3 - 4 | 301 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |