Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129897.4 | NP_181864.1 | 4 - 5 | 819 | 61 | |
GenBank mRNA | BT024899.1 | - | 4 - 5 | 696 | 61 | |
L34413.1 | - | 4 - 5 | 756 | 61 | ||
U31806.1 | - | 4 - 5 | 753 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |