Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129953.3 | NP_181919.1 | 7 - 8 | 1059 | 68 | |
GenBank mRNA | AY044314.1 | - | 7 - 8 | 652 | 68 | |
AY086360.1 | - | 7 - 8 | 726 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |