Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ckl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148142.3 | NP_680447.1 | 1 - 2 | 551 | 79 | |
GenBank mRNA | AK118919.1 | - | 1 - 2 | 482 | 79 | |
AY070458.1 | - | 1 - 2 | 389 | 79 | ||
AY091321.1 | - | 1 - 2 | 71 | 79 | ||
AY943855.1 | - | 1 - 2 | 71 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |