Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345714.1 | NP_001331432.1 | 7 - 8 | 1648 | 97 | |
NM_001345715.1 | NP_001331431.1 | 7 - 8 | 1648 | 97 | ||
NM_001345716.1 | NP_001331433.1 | 6 - 7 | 1745 | 97 | ||
NM_148163.3 | NP_680468.1 | 7 - 8 | 1727 | 97 | ||
GenBank mRNA | BT046193.1 | - | 6 - 7 | 1534 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |