Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_202562.5 | NP_974291.4 | 1 - 2 | 823 | 85 | |
GenBank mRNA | AY059802.1 | - | 1 - 2 | 613 | 85 | |
AY085937.1 | - | 1 - 2 | 660 | 85 | ||
BT001172.1 | - | 1 - 2 | 596 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |