Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084701.1 | NP_001078170.1 | 3 | 1187 | 125 | |
NM_180275.3 | NP_850606.1 | 2 | 1283 | 125 | ||
NM_202600.1 | NP_974329.1 | 1 | 1389 | 125 | ||
GenBank mRNA | AY080647.1 | - | 2 | 1182 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |