Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127036.5 | NP_179079.1 | 4 - 5 | 1445 | 155 | |
NM_179623.2 | NP_849954.1 | 4 - 5 | 1384 | 155 | ||
GenBank mRNA | AY062744.1 | - | 4 - 5 | 1323 | 155 | |
BT008390.1 | - | 4 - 5 | 1156 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |