Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127293.3 | NP_565415.1 | 4 - 5 | 415 | 163 | |
GenBank mRNA | AY085130.1 | - | 4 - 5 | 405 | 163 | |
BT025619.1 | - | 3 - 4 | 285 | 163 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |