Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202701.1 | NP_001189630.1 | 8 - 9 | 3083 | 173 | |
NM_128475.4 | NP_180482.1 | 8 - 9 | 3065 | 173 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |