Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRT2:1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100684.3 | NP_172288.1 | 2 - 3 | 1041 | 142 | |
GenBank mRNA | AF019748.1 | - | 2 - 3 | 969 | 142 | |
AF093754.1 | - | 2 - 3 | 990 | 142 | ||
Z97058.1 | - | 989 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Inventoried Probe spans exons |