Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP79F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101507.3 | NP_563996.2 | 1 - 2 | 1151 | 133 | |
NM_202111.2 | NP_973840.1 | 1 - 2 | 1026 | 133 | ||
GenBank mRNA | AY050890.1 | - | 1 - 2 | 1026 | 133 | |
AY114074.1 | - | 1 - 2 | 971 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |