Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABA1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101553.3 | NP_173136.1 | 1 - 2 | 388 | 151 | |
GenBank mRNA | AK227865.1 | - | 1 - 2 | 295 | 151 | |
BT006406.1 | - | 1 - 2 | 223 | 151 | ||
L18883.1 | - | 1 - 2 | 277 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |