Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BES1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084093.1 | NP_001077562.1 | 2 - 3 | 310 | 148 | |
NM_101792.4 | NP_564081.1 | 1 - 2 | 402 | 148 | ||
NM_202134.1 | NP_973863.1 | 2 - 3 | 470 | 148 | ||
NM_202135.3 | NP_973864.1 | 2 - 3 | 507 | 148 | ||
NM_202136.1 | NP_973865.1 | 2 - 3 | 473 | 148 | ||
GenBank mRNA | AY065041.1 | - | 1 - 2 | 396 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |