Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105298.4 | NP_176801.1 | 10 - 11 | 1327 | 148 | |
NM_179517.2 | NP_849848.1 | 10 - 11 | 1196 | 148 | ||
GenBank mRNA | AY045698.1 | - | 10 - 11 | 1197 | 148 | |
BT002684.1 | - | 10 - 11 | 1156 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |