Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105298.4 | NP_176801.1 | 11 - 12 | 1427 | 79 | |
NM_179517.2 | NP_849848.1 | 11 - 12 | 1296 | 79 | ||
GenBank mRNA | AY045698.1 | - | 11 - 12 | 1297 | 79 | |
BT002684.1 | - | 11 - 12 | 1256 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |