Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERD15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202798.1 | NP_001189727.1 | 2 - 3 | 639 | 101 | |
NM_129706.4 | NP_181674.1 | 2 - 3 | 620 | 101 | ||
NM_180019.3 | NP_850350.1 | 2 - 3 | 606 | 101 | ||
NM_201929.2 | NP_973658.1 | 1 - 2 | 849 | 101 | ||
GenBank mRNA | AF372909.1 | - | 2 - 3 | 475 | 101 | |
AK317136.1 | - | 2 - 3 | 413 | 101 | ||
AY056399.1 | - | 2 - 3 | 475 | 101 | ||
AY065120.1 | - | 2 - 3 | 478 | 101 | ||
AY081636.1 | - | 1 - 2 | 345 | 101 | ||
BT002663.1 | - | 1 - 2 | 345 | 101 | ||
D30719.1 | - | 2 - 3 | 463 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |