Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP76C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337143.1 | NP_001325389.1 | 1 - 2 | 414 | 142 | |
NM_180108.2 | NP_850439.1 | 1 - 2 | 716 | 142 | ||
NM_180109.3 | NP_850440.1 | 1 - 2 | 414 | 142 | ||
GenBank mRNA | AY078939.1 | - | 1 - 2 | 548 | 142 | |
AY124874.1 | - | 1 - 2 | 394 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |