Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111692.3 | NP_187470.1 | 2 - 3 | 522 | 145 | |
NM_180210.2 | NP_850541.1 | 2 - 3 | 742 | 145 | ||
GenBank mRNA | AK226814.1 | - | 600 | 145 | ||
AY034933.1 | - | 2 - 3 | 585 | 145 | ||
AY042814.1 | - | 2 - 3 | 593 | 145 | ||
AY054248.1 | - | 2 - 3 | 582 | 145 | ||
AY074529.1 | - | 1 - 2 | 472 | 145 | ||
BT000381.1 | - | 1 - 2 | 472 | 145 | ||
BT002387.1 | - | 2 - 3 | 585 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |