Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alpha-ADR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343762.1 | NP_001330970.1 | 13 - 14 | 1696 | 108 | |
NM_001343763.1 | NP_001330971.1 | 14 - 15 | 1587 | 108 | ||
NM_001343764.1 | NP_001330969.1 | 14 - 15 | 1565 | 108 | ||
NM_122183.3 | NP_197669.1 | 14 - 15 | 1550 | 108 | ||
NM_180726.2 | NP_851057.1 | 13 - 14 | 1696 | 108 | ||
NM_180727.2 | NP_851058.1 | 14 - 15 | 1530 | 108 | ||
GenBank mRNA | AY099568.1 | - | 13 - 14 | 1618 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |