Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201863.2 | NP_973592.1 | 1 - 2 | 882 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |