Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113642.4 | NP_189364.1 | 1 - 2 | 408 | 140 | |
NM_202640.2 | NP_974369.1 | 1 - 2 | 398 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |