Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSP91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036228.2 | NP_001031305.1 | 4 - 5 | 1686 | 174 | |
NM_106642.5 | NP_178111.1 | 4 - 5 | 1728 | 174 | ||
GenBank mRNA | AK226836.1 | - | 4 - 5 | 1639 | 174 | |
AY079015.1 | - | 4 - 5 | 1638 | 174 | ||
BT002625.1 | - | 3 - 4 | 1488 | 174 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |