Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sks14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104432.5 | NP_564697.1 | 1 - 2 | 372 | 129 | |
GenBank mRNA | AJ249211.1 | - | 1 - 2 | 378 | 129 | |
AK118598.1 | - | 1 - 2 | 352 | 129 | ||
BT005910.1 | - | 1 - 2 | 243 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |