Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111470.2 | NP_187247.1 | 2 - 3 | 1239 | 134 | |
GenBank mRNA | AJ344337.1 | - | 2 - 3 | 1078 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |