Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP71B19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113524.5 | NP_189248.1 | 1 - 2 | 977 | 125 | |
GenBank mRNA | AK118446.1 | - | 1 - 2 | 910 | 125 | |
BT005891.1 | - | 1 - 2 | 891 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |