Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABA1i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102623.2 | NP_174177.1 | 1 - 2 | 398 | 117 | |
GenBank mRNA | DQ056467.1 | - | 1 - 2 | 224 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |