Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT78D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121709.2 | NP_197205.1 | 1 - 2 | 668 | 170 | |
GenBank mRNA | AY088168.1 | - | 1 - 2 | 513 | 170 | |
KJ138683.1 | - | 1 - 2 | 498 | 170 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |