Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084778.1 | NP_001078247.1 | 2 - 3 | 442 | 157 | |
NM_114470.3 | NP_190187.1 | 2 - 3 | 440 | 157 | ||
GenBank mRNA | AY035154.1 | - | 2 - 3 | 411 | 157 | |
AY062940.1 | - | 1 - 2 | 273 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |