Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABA2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339276.1 | NP_001327707.1 | 2 - 3 | 373 | 134 | |
NM_114550.4 | NP_190267.1 | 2 - 3 | 502 | 134 | ||
GenBank mRNA | AY065380.1 | - | 2 - 3 | 353 | 134 | |
AY087450.1 | - | 2 - 3 | 377 | 134 | ||
AY096545.1 | - | 1 - 2 | 163 | 134 | ||
Y08904.1 | - | 333 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |