Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340028.1 | NP_001327770.1 | 2 - 3 | 2409 | 164 | |
NM_001340029.1 | NP_001327769.1 | 1 - 2 | 2755 | 164 | ||
NM_115879.7 | NP_191575.2 | 2 - 3 | 2588 | 164 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |