Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160744.1 | NP_001154216.1 | 1 - 2 | 667 | 151 | |
NM_116969.3 | NP_567344.1 | 1 - 2 | 667 | 151 | ||
GenBank mRNA | AF412067.1 | - | 1 - 2 | 578 | 151 | |
AY065064.1 | - | 1 - 2 | 611 | 151 | ||
AY086143.1 | - | 1 - 2 | 548 | 151 | ||
BT024489.1 | - | 1 - 2 | 490 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |