Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLDGAMMA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340738.1 | NP_001329644.1 | 6 - 7 | 1957 | 158 | |
NM_117254.3 | NP_192921.1 | 5 - 6 | 2047 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |