Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDA36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340891.1 | NP_001328680.1 | 1 - 2 | 1078 | 107 | |
NM_117463.3 | NP_193125.1 | 1 - 2 | 1078 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |