Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDA36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117463.3 | NP_193125.1 | 2 - 3 | 1187 | 173 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |