Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122009.4 | NP_197502.1 | 3 - 4 | 383 | 111 | |
GenBank mRNA | AF370337.1 | - | 3 - 4 | 193 | 111 | |
AK226813.1 | - | 197 | 111 | |||
AY062997.1 | - | 2 - 3 | 135 | 111 | ||
BT000424.1 | - | 3 - 4 | 200 | 111 | ||
BT000659.1 | - | 3 - 4 | 196 | 111 | ||
BT006264.1 | - | 2 - 3 | 135 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |