Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP71B13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122423.3 | NP_197896.1 | 2 - 3 | 1105 | 142 | |
GenBank mRNA | AK227044.1 | - | 2 - 3 | 998 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |