Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123489.4 | NP_198940.3 | 3 - 4 | 253 | 149 | |
GenBank mRNA | AY139996.1 | - | 3 - 4 | 246 | 149 | |
BT008361.1 | - | 3 - 4 | 199 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |