Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCS20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123743.4 | NP_199189.1 | 1 - 2 | 1167 | 174 | |
GenBank mRNA | AF424620.1 | - | 1 - 2 | 917 | 174 | |
AY150469.1 | - | 1 - 2 | 787 | 174 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |