Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHX25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125232.2 | NP_200654.1 | 1 - 2 | 297 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |