Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RD28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129274.4 | NP_181255.1 | 3 - 4 | 881 | 160 | |
GenBank mRNA | AY064029.1 | - | 3 - 4 | 804 | 160 | |
AY096701.1 | - | 3 - 4 | 737 | 160 | ||
D13254.1 | - | 3 - 4 | 787 | 160 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |