Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PP2A-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202803.2 | NP_001189732.1 | 2 - 3 | 338 | 156 | |
NM_001336978.1 | NP_001324021.1 | 2 - 3 | 338 | 156 | ||
NM_001336979.1 | NP_001324022.1 | 2 - 3 | 338 | 156 | ||
NM_129811.4 | NP_565974.1 | 2 - 3 | 338 | 156 | ||
GenBank mRNA | BT010166.1 | - | 2 - 3 | 147 | 156 | |
M96841.1 | - | 2 - 3 | 335 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |