Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GH9B11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130020.3 | NP_181984.1 | 1 - 2 | 299 | 119 | |
GenBank mRNA | AK175889.1 | - | 1 - 2 | 277 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |